Gene Transcripts

Gene Symbol
Tecpr2
Entrez Gene ID
104859
Gene Name
tectonin beta-propeller repeat containing 2
Gene Aliases
4930573I19Rik, AI604920, mKIAA0297
Chromosome Location
Chr. 12: 110889259 - 110972394 on Build GRCm38
UniGene ID
Mm.35583
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081057.2 NP_001074526.1 19 4104
NM_001289510.1 NP_001276439.1 18 4052
NM_175336.3 NP_780545.2 19 4214
GenBank mRNA AK029810.1 - 1 29
AK129113.1 - 18 4056
AK147592.1 - 18 4052
BC041778.1 - 16 3696
BC138354.1 - 18 4047 8861374
BC145250.1 - 18 4157

View Full Product Details