Gene Transcripts

Gene Symbol
Akr1c14
Entrez Gene ID
105387
Gene Name
aldo-keto reductase family 1, member C14
Gene Aliases
9030611N15Rik, AW557061
Chromosome Location
Chr. 13: 4048999 - 4090560 on Build GRCm38
UniGene ID
Mm.26838
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134072.1 NP_598833.1 3 331
GenBank mRNA AK033537.1 - 3 349
AK136528.1 - 3 348
AY730283.1 - 3 293
BC013482.1 - 3 331 4240772
BC026628.1 - 3 330 4983237

View Full Product Details