Gene Transcripts

Gene Symbol
Oxsr1
Entrez Gene ID
108737
Gene Name
oxidative-stress responsive 1
Gene Aliases
2210022N24Rik, 2810422B09Rik, AI462649, AW209236, Osr1, mKIAA1101
Chromosome Location
Chr. 9: 119238432 - 119322710 on Build GRCm38
UniGene ID
Mm.293565
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133985.2 NP_598746.2 3, 4 654
GenBank mRNA AK031790.1 - 2, 3 151
AK075837.1 - 2, 3 162
AK129291.1 - 3, 4 648
BC046453.1 - 4, 5 678
BC060645.1 - 3, 4 637 6852021

View Full Product Details