Gene Transcripts

Gene Symbol
DHRS2
Entrez Gene ID
10202
Gene Name
dehydrogenase/reductase 2
Gene Aliases
HEP27, SDR25C1
Chromosome Location
Chr. 14: 23630115 - 23645639 on Build GRCh38
UniGene ID
Hs.272499
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005794.3 NP_005785.1 9 1569
NM_182908.4 NP_878912.1 9 1580
XM_005267249.1 XP_005267306.1 9 1281
XM_006720001.3 XP_006720064.1 9 1244
XM_011536338.1 XP_011534640.1 8 1210
XM_011536339.2 XP_011534641.1 8 1060
GenBank mRNA AB209653.1 - 7 5211
AK000345.1 - 1 2488
AK092834.1 - 5 2980
AK222857.1 - 9 1569
AK309945.1 - 9 3056
BC002786.2 - 9 1571 3614599
BC007339.2 - 8 1095

View Full Product Details