Gene Transcripts

Gene Symbol
CYP11A1
Entrez Gene ID
1583
Gene Name
cytochrome P450 family 11 subfamily A member 1
Gene Aliases
CYP11A, CYPXIA1, P450SCC
Chromosome Location
Chr. 15: 74337762 - 74367740 on Build GRCh38
UniGene ID
Hs.303980
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000781.2 NP_000772.2 1 346
GenBank mRNA AK225715.1 - 1 252
AK292300.1 - 1 252
AK293211.1 - 1 252
AK300178.1 - 1 252
AK310362.1 - 1 252
BC032329.1 - 1 235 5165263
BX398579.2 - 1 346
KU178011.1 - 1 191
KU178012.1 - 1 191
KU178013.1 - 1 191
KU178014.1 - 1 191
M14565.1 - 1 235

View Full Product Details