Gene Transcripts

Gene Symbol
SPATA17
Entrez Gene ID
128153
Gene Name
spermatogenesis associated 17
Gene Aliases
IQCH, MSRG-11, MSRG11
Chromosome Location
Chr. 1: 217631324 - 217868151 on Build GRCh38
UniGene ID
Hs.171130
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138796.3 NP_620151.1 8 862
XM_005273052.1 XP_005273109.1 8 844
XM_006711165.3 XP_006711228.1 7 802
GenBank mRNA AK098591.1 - 8 842
AK310291.1 - 11 1184
AY963797.1 - 8 865
BC014608.2 - 8 833 4824429

Gene Symbol
SPATA17-AS1
Entrez Gene ID
103752555
Gene Name
SPATA17 antisense RNA 1
Gene Aliases
Chromosome Location
Chr. 1: 217781198 - 217785120 on Build GRCh38
UniGene ID
Hs.602803
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AI025201.1 - 2 225

View Full Product Details