Gene Transcripts

Gene Symbol
IGSF22
Entrez Gene ID
283284
Gene Name
immunoglobulin superfamily member 22
Gene Aliases
IGFN2
Chromosome Location
Chr. 11: 18704305 - 18726939 on Build GRCh38
UniGene ID
Hs.434152
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173588.3 NP_775859.3 2, 3 293
XM_017017608.1 XP_016873097.1 1, 2 1111
GenBank mRNA AK095113.1 - 2, 3 293
BC115396.1 - 2, 3 244
BC115397.1 - 2, 3 244 40068579
DA338373.1 - 2, 3 251

View Full Product Details