Gene Transcripts

Gene Symbol
TEPP
Entrez Gene ID
374739
Gene Name
testis, prostate and placenta expressed
Gene Aliases
Chromosome Location
Chr. 16: 57976435 - 57988116 on Build GRCh38
UniGene ID
Hs.132976
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199046.2 NP_950247.2 7, 8 905
NM_199456.2 NP_955535.2 7, 8 824
GenBank mRNA AY363222.1 - 7, 8 824
AY363223.1 - 7, 8 905
BC104458.1 - 7, 8 815 40030655
BC104459.1 - 7, 8 815 40030657

View Full Product Details