Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017864.3 | NP_060334.2 | 17 | 2235 | |
NR_073444.1 | 17 | 2235 | |||
NR_073445.1 | 17 | 2204 | |||
XM_017013616.1 | XP_016869105.1 | 17 | 2235 | ||
XM_017013618.1 | XP_016869107.1 | 14 | 1788 | ||
XM_017013619.1 | XP_016869108.1 | 17 | 2194 | ||
GenBank mRNA | AB161944.1 | - | 17 | 2219 | |
AK000537.1 | - | 7 | 789 | ||
AK001403.1 | - | 10 | 1175 | ||
AK091278.1 | - | 16 | 1972 | ||
AK225533.1 | - | 7 | 789 | ||
BC050536.1 | - | 9 | 1023 | ||
BC064915.1 | - | 9 | 1064 | ||
BC136754.1 | - | 17 | 2149 | 9020744 | |
BC144396.1 | - | 17 | 2149 | ||
BC144397.1 | - | 17 | 2118 | ||
BX538203.1 | - | 13 | 1558 |