Gene Transcripts

Gene Symbol
CXorf57
Entrez Gene ID
55086
Gene Name
chromosome X open reading frame 57
Gene Aliases
Chromosome Location
Chr. X: 106611930 - 106679443 on Build GRCh38
UniGene ID
Hs.274267
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184782.1 NP_001171711.1 1 386
NM_018015.5 NP_060485.4 1 386
GenBank mRNA AK001040.1 - 1 341
AK291730.1 - 1 335
BC011483.1 - 1 307 3453700
BC033709.1 - 1 298 3910777
BC054032.2 - 1 274
BC070110.1 - 1 337 5264812
DB065928.1 - 1 389

View Full Product Details