Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021637.2 | NP_067650.1 | 2 | 617 | |
GenBank mRNA | AK024146.1 | - | 2 | 590 | |
AK223240.1 | - | 2 | 585 | ||
AK297745.1 | - | 3 | 459 | ||
BC050273.1 | - | 2 | 582 | 4801325 | |
BC078658.1 | - | 2 | 582 | 5261160 |