Gene Transcripts

Gene Symbol
C9orf24
Entrez Gene ID
84688
Gene Name
chromosome 9 open reading frame 24
Gene Aliases
CBE1, NYD-SP22, SMRP1, bA573M23.4
Chromosome Location
Chr. 9: 34379019 - 34400833 on Build GRCh38
UniGene ID
Hs.50334
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252195.1 NP_001239124.1 5 562
NM_032596.3 NP_115985.2 7 1096
NM_147168.1 NP_671697.1 4 638
NM_147169.2 NP_671698.1 4 484
XM_005251616.4 XP_005251673.1 7 4083
XM_005251618.4 XP_005251675.1 5 630
XM_011518055.2 XP_011516357.1 5 904
GenBank mRNA AF367474.1 - 4 423
AK058087.1 - 7 1034
AK094751.1 - 2 2260
AK225981.1 - 4 556
BC022084.1 - 7 1081 4825995
BC029484.1 - 7 1096 5271301
CB854584.1 - 4 607

View Full Product Details