Gene Transcripts

Gene Symbol
PSME4
Entrez Gene ID
23198
Gene Name
proteasome activator subunit 4
Gene Aliases
PA200
Chromosome Location
Chr. 2: 53864067 - 53970840 on Build GRCh38
UniGene ID
Hs.413801
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014614.2 NP_055429.2 5 691
XM_006711969.1 XP_006712032.1 3 207
XM_011532705.2 XP_011531007.1 6 715
XM_011532706.2 XP_011531008.1 6 715
XM_011532707.2 XP_011531009.1 5 649
XM_011532709.2 XP_011531011.1 6 715
GenBank mRNA AB621805.1 - 4 808
AL599601.1 - 5 564
AY894754.1 - 5 691
AY894756.1 - 5 635
BC112169.1 - 4 327 8327637
BC113668.1 - 4 327 8322720
BC143739.1 - 4 327
D38521.1 - 5 501

View Full Product Details