Gene Transcripts

Gene Symbol
CPNE6
Entrez Gene ID
9362
Gene Name
copine 6
Gene Aliases
Chromosome Location
Chr. 14: 24070837 - 24078100 on Build GRCh38
UniGene ID
Hs.6132
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001280558.1 NP_001267487.1 7 868
NM_006032.3 NP_006023.1 6 856
XM_005268216.1 XP_005268273.1 7 812
XM_005268217.1 XP_005268274.1 5 506
GenBank mRNA AB009288.1 - 6 656
AK125195.1 - 5 1418
AK222675.1 - 6 616
AK293655.1 - 7 868
AK294460.1 - 5 771
AK314182.1 - 6 669
AL137688.1 - 3 2231
BC018046.1 - 6 662 4799650

View Full Product Details