Gene Transcripts

Gene Symbol
ITGA10
Entrez Gene ID
8515
Gene Name
integrin subunit alpha 10
Gene Aliases
PRO827
Chromosome Location
Chr. 1: 145891208 - 145910189 on Build GRCh38
UniGene ID
Hs.158237
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303040.1 NP_001289969.1 5 632
NM_001303041.1 NP_001289970.1 4 520
NM_003637.4 NP_003628.2 8 949
XM_005277436.3 XP_005277493.1 7 700
XM_011510083.2 XP_011508385.1 4 380
XM_017002622.1 XP_016858111.1 6 906
XM_017002623.1 XP_016858112.1 6 906
XM_017002624.1 XP_016858113.1 6 906
XM_017002625.1 XP_016858114.1 6 906
XM_017002626.1 XP_016858115.1 4 663
XM_017002627.1 XP_016858116.1 6 906
XM_017002628.1 XP_016858117.1 1, 2 356
GenBank mRNA AF074015.1 - 8 795
AF112345.1 - 8 850
AK304155.1 - 5 488
AK304183.1 - 7 779
AK314255.1 - 8 798
BC140831.1 - 4 395 9021699
BC144637.1 - 4 395 9053168

View Full Product Details