Gene Transcripts

Gene Symbol
ESYT1
Entrez Gene ID
23344
Gene Name
extended synaptotagmin 1
Gene Aliases
FAM62A, MBC2
Chromosome Location
Chr. 12: 56128202 - 56144676 on Build GRCh38
UniGene ID
Hs.632729
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184796.1 NP_001171725.1 12 1460
NM_015292.2 NP_056107.1 12 1460
GenBank mRNA AB018290.1 - 12 1217
AK022790.1 - 12 1411
AK025463.1 - 8 665
AK128730.1 - 11 1264
AK225035.1 - 12 1395
AK290337.1 - 12 1395
AL050134.1 - 4 208
BC004998.1 - 12 1392 2958094
DQ993200.1 - 12 1342

View Full Product Details