Gene Transcripts

Gene Symbol
TSR1
Entrez Gene ID
55720
Gene Name
TSR1, ribosome maturation factor
Gene Aliases
Chromosome Location
Chr. 17: 2322678 - 2337384 on Build GRCh38
UniGene ID
Hs.388170
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018128.4 NP_060598.3 5 1777
GenBank mRNA AB037822.1 - 5 1900
AK001396.1 - 3 419
AK026565.1 - 5 850
BC019090.2 - 4 685
BC110851.1 - 5 1355 5786818
BC126110.1 - 5 1325 8991826

View Full Product Details