Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001204477.1 | NP_001191406.1 | 4 | 1134 | |
GenBank mRNA | AK093993.1 | - | 2 | 878 | |
AK172733.1 | - | 1 | 271 | ||
DB105410.1 | - | 1 | 544 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001204478.1 | NP_001191407.1 | 7 | 1545 | |
NR_037924.1 | 6 | 1400 | |||
GenBank mRNA | AB074181.1 | - | 7 | 1475 |