Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001292048.1 | NP_001278977.1 | 6 | 1877 | |
NM_004162.4 | NP_004153.2 | 6 | 1919 | ||
GenBank mRNA | AF464088.1 | - | 6 | 1760 | |
BC001267.1 | - | 6 | 1549 | 3463669 | |
BC018288.1 | - | 6 | 1603 | 3454268 |