Gene Transcripts

Gene Symbol
MOK
Entrez Gene ID
5891
Gene Name
MOK protein kinase
Gene Aliases
RAGE, RAGE-1, RAGE1, STK30
Chromosome Location
Chr. 14: 102224450 - 102305200 on Build GRCh38
UniGene ID
Hs.104119
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001272011.1 NP_001258940.1 7, 8 830
NM_014226.2 NP_055041.1 8, 9 920
NR_073540.1 2, 3 268
NR_073541.1 3, 4 315
NR_073542.1 2, 3 268
XM_006720225.2 XP_006720288.1 8, 9 920
XM_006720226.2 XP_006720289.1 8, 9 920
XM_011537052.1 XP_011535354.1 8, 9 920
XM_011537053.2 XP_011535355.1 2, 3 947
XM_017021549.1 XP_016877038.1 8, 9 917
XM_017021550.1 XP_016877039.1 7, 8 816
XM_017021551.1 XP_016877040.1 9, 10 860
XM_017021552.1 XP_016877041.1 8, 9 958
XM_017021553.1 XP_016877042.1 6, 7 755
XM_017021554.1 XP_016877043.1 6, 7 754
XM_017021555.1 XP_016877044.1 8, 9 917
XM_017021557.1 XP_016877046.1 7, 8 820
XM_017021558.1 XP_016877047.1 7, 8 783
XM_017021559.1 XP_016877048.1 7, 8 909
GenBank mRNA AB022694.1 - 8, 9 914
AK302349.1 - 7, 8 815
AK312778.1 - 8, 9 911
BC026069.1 - 7, 8 1331 4838296
BC043179.1 - 6, 7 1676 5287879
U46192.1 - 2, 3 258
U46193.1 - 3, 4 315
U46194.1 - 2, 3 254

View Full Product Details