Gene Transcripts

Gene Symbol
MLANA
Entrez Gene ID
2315
Gene Name
melan-A
Gene Aliases
MART-1, MART1
Chromosome Location
Chr. 9: 5890909 - 5909822 on Build GRCh38
UniGene ID
Hs.154069
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005511.1 NP_005502.1 5 1400
GenBank mRNA U06452.1 - 5 1400

View Full Product Details