Gene Transcripts

Gene Symbol
AFM
Entrez Gene ID
173
Gene Name
afamin
Gene Aliases
ALB2, ALBA, ALF
Chromosome Location
Chr. 4: 73481740 - 73504001 on Build GRCh38
UniGene ID
Hs.168718
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001133.2 NP_001124.1 9 1131
XM_017007842.1 XP_016863331.1 9 1136
XM_017007843.1 XP_016863332.1 9 1136
XM_017007844.1 XP_016863333.1 9 1136
GenBank mRNA AK290556.1 - 9 1131
BC109020.1 - 9 1129 40006696
BC109021.1 - 9 1129 40006697

View Full Product Details