Gene Transcripts
- Gene Symbol
- ACIN1
- Entrez Gene ID
- 22985
- Gene Name
- apoptotic chromatin condensation inducer 1
- Gene Aliases
- ACINUS, ACN, fSAP152
- Chromosome Location
- Chr. 14: 23058564 - 23095614 on Build GRCh38
- UniGene ID
- Hs.124490
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001164814.1 |
NP_001158286.1
|
2, 3 |
702 |
|
NM_001164815.1 |
NP_001158287.1
|
2, 3 |
702 |
|
NM_014977.3 |
NP_055792.1
|
2, 3 |
702 |
|
XM_005267415.3 |
XP_005267472.1
|
2, 3 |
506 |
|
XM_005267416.3 |
XP_005267473.1
|
2, 3 |
506 |
|
XM_006720081.3 |
XP_006720144.1
|
2, 3 |
506 |
|
XM_011536570.2 |
XP_011534872.1
|
2, 3 |
506 |
|
GenBank mRNA |
AB014570.2 |
- |
2, 3 |
211 |
|
AF124726.1 |
- |
2, 3 |
701 |
|
AK299034.1 |
- |
2, 3 |
207 |
|
BC140805.1 |
- |
2, 3 |
471 |
9021673 |
View Full Product Details