Gene Transcripts

Gene Symbol
RBM19
Entrez Gene ID
9904
Gene Name
RNA binding motif protein 19
Gene Aliases
Chromosome Location
Chr. 12: 113816738 - 113966371 on Build GRCh38
UniGene ID
Hs.7482
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146698.1 NP_001140170.1 9 1197
NM_001146699.1 NP_001140171.1 9 1197
NM_016196.3 NP_057280.2 9 1197
GenBank mRNA AB014582.1 - 9 1132
AK291444.1 - 9 1148
AK314606.1 - 9 1197
AL117547.1 - 9 1138
BC004289.1 - 9 1136 3619715
BC006137.1 - 9 1136 3677625

View Full Product Details