Gene Transcripts

Gene Symbol
Clpb
Entrez Gene ID
20480
Gene Name
ClpB caseinolytic peptidase B
Gene Aliases
AL118244, Skd3
Chromosome Location
Chr. 7: 101663595 - 101790168 on Build GRCm38
UniGene ID
Mm.3990
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009191.3 NP_033217.1 10, 11 1218
GenBank mRNA AK032421.1 - 1213
AK138593.1 - 10, 11 1256
AK141383.1 - 10, 11 1227
AK145641.1 - 10, 11 1210
AK147772.1 - 10, 11 939
AK154577.1 - 10, 11 1239
AK159314.1 - 10, 11 1259
AK186387.1 - 1, 2 44
BC048175.1 - 10, 11 1218 6825472
U09874.1 - 10, 11 1220

View Full Product Details