Gene Transcripts

Gene Symbol
Kirrel2
Entrez Gene ID
243911
Gene Name
kin of IRRE like 2 (Drosophila)
Gene Aliases
C330019F22Rik, NEPH3, NLG1
Chromosome Location
Chr. 7: 30447530 - 30460680 on Build GRCm38
UniGene ID
Mm.113908
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172898.3 NP_766486.1 15 3093
GenBank mRNA AK049284.1 - 15 3094
AK083069.1 - 13 3643
BC052773.1 - 15 2556 6314924

View Full Product Details