Gene Transcripts

Gene Symbol
Ralgps1
Entrez Gene ID
241308
Gene Name
Ral GEF with PH domain and SH3 binding motif 1
Gene Aliases
5830418G11Rik, AI853783, AI854138, RALGEF2, RALGPS1A, RalGEF 2, mKIAA0351
Chromosome Location
Chr. 2: 33133419 - 33371511 on Build GRCm38
UniGene ID
Mm.274249
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290570.1 NP_001277499.1 4, 5 546
NM_001290572.1 NP_001277501.1 5, 6 597
NM_175211.5 NP_780420.1 5, 6 597
GenBank mRNA AK076767.1 - 5, 6 582
AK129123.1 - 4, 5 513
AK170248.1 - 5, 6 544
BC072656.1 - 5, 6 532 30536315

View Full Product Details