Gene Transcripts

Gene Symbol
Coro2b
Entrez Gene ID
235431
Gene Name
coronin, actin binding protein, 2B
Gene Aliases
CLIPINC, E130012P22Rik
Chromosome Location
Chr. 9: 62419490 - 62537044 on Build GRCm38
UniGene ID
Mm.335229
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175484.3 NP_780693.2 12 2537
GenBank mRNA AK087414.1 - 12 2538
AK194455.1 - 1 155
BC031897.1 - 1 1219
BC062649.1 - 12 2269 6416110

View Full Product Details