Gene Transcripts

Gene Symbol
Gpr150
Entrez Gene ID
238725
Gene Name
G protein-coupled receptor 150
Gene Aliases
C030001A19Rik, PGR11
Chromosome Location
Chr. 13: 76054851 - 76056996 on Build GRCm38
UniGene ID
Mm.167172
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175495.2 NP_780704.1 1 2079
GenBank mRNA AK047609.1 - 1 2079

View Full Product Details