Gene Transcripts

Gene Symbol
Pnma2
Entrez Gene ID
239157
Gene Name
paraneoplastic antigen MA2
Gene Aliases
A830049P17Rik, mKIAA0883
Chromosome Location
Chr. 14: 66900682 - 66920063 on Build GRCm38
UniGene ID
Mm.44358
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175498.4 NP_780707.1 3 1571
GenBank mRNA AK043718.1 - 3 1519
AK043910.1 - 2 1441
AK220357.1 - 1526
BC065116.1 - 2 1402 30618233

View Full Product Details