Gene Transcripts

Gene Symbol
Pias2
Entrez Gene ID
17344
Gene Name
protein inhibitor of activated STAT 2
Gene Aliases
6330408K17Rik, AI462206, ARIP3, AU018068, Dib, Miz1, PIASxalpha, PIASxb, PIASxbeta, SIZ2
Chromosome Location
Chr. 18: 77065208 - 77155710 on Build GRCm38
UniGene ID
Mm.6370
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164167.1 NP_001157639.1 12 1775
NM_001164168.1 NP_001157640.1 12 1791
NM_001164169.1 NP_001157641.1 12 1775
NM_001164170.1 NP_001157642.1 12 1791
NM_008602.4 NP_032628.3 12 1775
GenBank mRNA AF039567.1 - 11 1220
AF201391.1 - 12 1702
AK014871.1 - 12 1766
AK029716.1 - 12 1776
AK032463.1 - 2 210
AK078097.1 - 7 842
AK078813.1 - 12 1745
AK086653.1 - 12 1707
AK149142.1 - 13 1814
BC005596.1 - 12 1652 3969546
BC034711.1 - 12 1781 2631738

View Full Product Details