Gene Transcripts

Gene Symbol
Lmnb2
Entrez Gene ID
16907
Gene Name
lamin B2
Gene Aliases
Chromosome Location
Chr. 10: 80901363 - 80918245 on Build GRCm38
UniGene ID
Mm.7362
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010722.5 NP_034852.2 12 3294
GenBank mRNA AK012232.1 - 1 891
BC042430.1 - 12 3299 3674002
BC051985.1 - 12 3270 5695459
X54098.1 - 12 3243

View Full Product Details