Gene Transcripts

Gene Symbol
Arc
Entrez Gene ID
11838
Gene Name
activity regulated cytoskeletal-associated protein
Gene Aliases
Arc3.1, C86064, arg3.1, mArc
Chromosome Location
Chr. 15: 74669081 - 74672570 on Build GRCm38
UniGene ID
Mm.491310
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276684.1 NP_001263613.1 1 1763
NM_018790.3 NP_061260.1 1 1763
GenBank mRNA AF162777.1 - 1 1754
AK157822.1 - 1 1764
BC023127.1 - 1 1752 3498057

View Full Product Details