Gene Transcripts

Gene Symbol
Anapc7
Entrez Gene ID
56317
Gene Name
anaphase promoting complex subunit 7
Gene Aliases
APC7, AW545589
Chromosome Location
Chr. 5: 122422444 - 122444912 on Build GRCm38
UniGene ID
Mm.37341
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019805.4 NP_062779.3 9 1421
GenBank mRNA AF076607.2 - 9 1385
AK030775.1 - 9 1386
AK049968.1 - 9 1360
AK137286.1 - 9 1385
AK150764.1 - 9 1385
AK150891.1 - 9 1385
AK153114.1 - 9 1385
AK159523.1 - 9 1353
AK164202.1 - 9 1390
BC006635.1 - 9 1374 3482261

View Full Product Details