Gene Transcripts
- Gene Symbol
- MLANA
- Entrez Gene ID
- 2315
- Gene Name
- melan-A
- Gene Aliases
- MART-1, MART1
- Chromosome Location
- Chr. 9: 5890909 - 5909822 on Build GRCh38
- UniGene ID
- Hs.154069
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005511.1 |
NP_005502.1
|
5 |
1323 |
|
GenBank mRNA |
U06452.1 |
- |
5 |
1323 |
|
View Full Product Details