Gene Transcripts

Gene Symbol
Olfr715
Entrez Gene ID
258776
Gene Name
olfactory receptor 715
Gene Aliases
MOR260-1
Chromosome Location
Chr. 7: 107128352 - 107129482 on Build GRCm38
UniGene ID
Mm.223207
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146780.2 NP_666991.1 1 483
GenBank mRNA BC139445.1 - 1 484
BC139446.1 - 1 484 8862468

Gene Symbol
Gm10081
Entrez Gene ID
384732
Gene Name
predicted gene 10081
Gene Aliases
EG384732, ENSMUSG00000060503
Chromosome Location
Chr. 7: 107105912 - 107106859 on Build GRCm38
UniGene ID
Mm.457863
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162940.1 NP_001156412.1 1 392
GenBank mRNA BC139756.1 - 1 383

View Full Product Details