Gene Transcripts

Gene Symbol
Plin4
Entrez Gene ID
57435
Gene Name
perilipin 4
Gene Aliases
S3-12, mKIAA1881
Chromosome Location
Chr. 17: 56100587 - 56109947 on Build GRCm38
UniGene ID
Mm.12966
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020568.3 NP_065593.2 5 589
GenBank mRNA AF064748.1 - 5 600
AK173286.1 - 5 509

View Full Product Details