Gene Transcripts

Gene Symbol
Tmem215
Entrez Gene ID
320500
Gene Name
transmembrane protein 215
Gene Aliases
A930001M12Rik
Chromosome Location
Chr. 4: 40472180 - 40475653 on Build GRCm38
UniGene ID
Mm.156365
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166009.1 NP_001159481.1 2 2005
NM_177175.4 NP_796149.2 2 2947
GenBank mRNA AK044217.1 - 2 2949
AK044627.1 - 2 2949

View Full Product Details