Gene Transcripts
- Gene Symbol
- CPNE6
- Entrez Gene ID
- 9362
- Gene Name
- copine 6
- Gene Aliases
- Chromosome Location
- Chr. 14: 24070837 - 24078100 on Build GRCh38
- UniGene ID
- Hs.6132
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001280558.1 |
NP_001267487.1
|
6, 7 |
817 |
|
NM_006032.3 |
NP_006023.1
|
5, 6 |
805 |
|
XM_005268216.1 |
XP_005268273.1
|
6, 7 |
761 |
|
XM_005268217.1 |
XP_005268274.1
|
4, 5 |
455 |
|
GenBank mRNA |
AB009288.1 |
- |
5, 6 |
605 |
|
AK125195.1 |
- |
4, 5 |
1367 |
|
AK222675.1 |
- |
5, 6 |
565 |
|
AK293655.1 |
- |
6, 7 |
817 |
|
AK294460.1 |
- |
4, 5 |
720 |
|
BC018046.1 |
- |
5, 6 |
611 |
4799650 |
View Full Product Details