Gene Transcripts

Gene Symbol
2310067B10Rik
Entrez Gene ID
71947
Gene Name
RIKEN cDNA 2310067B10 gene
Gene Aliases
Kiaa0195, mKIAA0195
Chromosome Location
Chr. 11: 115765433 - 115799033 on Build GRCm38
UniGene ID
Mm.23168
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028014.3 NP_082290.2 14 1741
GenBank mRNA AK155233.1 - 14 1616
BC053088.1 - 14 1656 5709104

View Full Product Details