Gene Transcripts

Gene Symbol
Taf7l
Entrez Gene ID
74469
Gene Name
TAF7-like RNA polymerase II, TATA box binding protein (TBP)-associated factor
Gene Aliases
4933438I11Rik, 50kDa, Taf2q
Chromosome Location
Chr. X: 134460116 - 134476526 on Build GRCm38
UniGene ID
Mm.103259
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028958.4 NP_083234.2 4 460
GenBank mRNA AF285574.1 - 4 264
AK017109.1 - 4 461
BC106854.2 - 4 252 40046501

View Full Product Details