Gene Transcripts

Gene Symbol
Usp46
Entrez Gene ID
69727
Gene Name
ubiquitin specific peptidase 46
Gene Aliases
1190009E20Rik, 2410018I08Rik, AI451644
Chromosome Location
Chr. 5: 73998726 - 74068411 on Build GRCm38
UniGene ID
Mm.473866
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177561.3 NP_808229.1 4 668
GenBank mRNA AK029604.1 - 3 520
AK145298.1 - 4 688
BC030888.1 - 2 435
BC039916.1 - 4 670 4950034

View Full Product Details