Gene Transcripts

Gene Symbol
Cxcl14
Entrez Gene ID
57266
Gene Name
chemokine (C-X-C motif) ligand 14
Gene Aliases
1110031L23Rik, 1200006I23Rik, AI414372, BMAC, BRAK, KS1, Kec, MIP-2g, MIP2gamma, NJAC, Scyb14, bolekine
Chromosome Location
Chr. 13: 56288643 - 56296792 on Build GRCm38
UniGene ID
Mm.30211
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019568.2 NP_062514.2 4 716
GenBank mRNA AB041614.1 - 4 716
AF144754.1 - 4 540
AF152377.1 - 4 466
AF192557.1 - 4 714
AF252873.1 - 4 714
AF352784.1 - 4 326
AF352785.1 - 4 326
AK004615.1 - 4 560
AK014351.1 - 4 506
AK076112.1 - 4 558
AK155356.1 - 4 554
AU035952.1 - 4 713
BC079661.1 - 3 356
BC147274.1 - 4 460 9056321
BC147275.1 - 4 460 9056329

View Full Product Details