Gene Transcripts

Gene Symbol
Asb3
Entrez Gene ID
65257
Gene Name
ankyrin repeat and SOCS box-containing 3
Gene Aliases
2400011J03Rik, Asb-3
Chromosome Location
Chr. 11: 30954194 - 31102704 on Build GRCm38
UniGene ID
Mm.440539
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023906.3 NP_076395.2 2 341
GenBank mRNA AF155354.1 - 1 156
AK010315.1 - 2 277
AK028489.1 - 2 334
AK038086.1 - 2 322
AK038897.1 - 2 292
AK143580.1 - 2 276
BB618690.1 - 2 343
BC023086.1 - 2 275 4947610

View Full Product Details