Gene Transcripts

Gene Symbol
Asb3
Entrez Gene ID
65257
Gene Name
ankyrin repeat and SOCS box-containing 3
Gene Aliases
2400011J03Rik, Asb-3
Chromosome Location
Chr. 11: 30954194 - 31102704 on Build GRCm38
UniGene ID
Mm.440539
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023906.3 NP_076395.2 4 563
GenBank mRNA AF155354.1 - 3 378
AK010315.1 - 4 499
AK028489.1 - 5 669
AK038086.1 - 4 544
AK143580.1 - 4 498
BB618690.1 - 4 565
BC023086.1 - 4 497 4947610

View Full Product Details