Gene Transcripts

Gene Symbol
Hiatl1
Entrez Gene ID
66631
Gene Name
hippocampus abundant transcript-like 1
Gene Aliases
5730414C17Rik, AU045608, AW539692
Chromosome Location
Chr. 13: 65065030 - 65113000 on Build GRCm38
UniGene ID
Mm.239196
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083901.1 NP_001077370.1 7, 8 997
NM_133680.3 NP_598441.3 6, 7 1094
GenBank mRNA AK146596.1 - 7, 8 990
BC033469.1 - 6, 7 1056 4951631
CN709077.1 - 3, 4 423

View Full Product Details