Gene Transcripts

Gene Symbol
Mga
Entrez Gene ID
29808
Gene Name
MAX gene associated
Gene Aliases
AV312082, C130042M01Rik, C80739, D030062C11Rik, Mad5
Chromosome Location
Chr. 2: 119871646 - 119970129 on Build GRCm38
UniGene ID
Mm.437053
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164274.1 NP_001157746.1 2 758
NM_013720.2 NP_038748.2 2 758
GenBank mRNA AF205935.1 - 1 572
AK012050.1 - 2 721
AK032570.1 - 2 739
AK046252.1 - 2 759
AK133775.1 - 2 975
AK139919.1 - 2 710
AK145491.1 - 2 707
AK162302.1 - 2 755
AK172962.1 - 2 681

View Full Product Details