Gene Transcripts

Gene Symbol
Coq5
Entrez Gene ID
52064
Gene Name
coenzyme Q5 homolog, methyltransferase (yeast)
Gene Aliases
1810014G04Rik, D5Ertd33e
Chromosome Location
Chr. 5: 115279666 - 115296974 on Build GRCm38
UniGene ID
Mm.290142
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026504.2 NP_080780.1 7 1686
GenBank mRNA AK007496.1 - 7 1687
AK009825.1 - 7 1673
AK045993.1 - 7 1693
AK050334.1 - 7 1693
AK180966.1 - 1 205
BC052345.1 - 7 1693 6585618

View Full Product Details