Gene Transcripts

Gene Symbol
Phka1
Entrez Gene ID
64561
Gene Name
phosphorylase kinase, alpha 1
Gene Aliases
Pcyt1b
Chromosome Location
Chr. X: 72377020 - 72515387 on Build Rnor_6.0
UniGene ID
Rn.218907
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022626.1 NP_072148.1 2 209
GenBank mRNA AF197561.1 - 2 209

View Full Product Details