Gene Transcripts

Gene Symbol
Stip1
Entrez Gene ID
192277
Gene Name
stress-induced phosphoprotein 1
Gene Aliases
Chromosome Location
Chr. 1: 222274133 - 222293139 on Build Rnor_6.0
UniGene ID
Rn.22614
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138911.3 NP_620266.1 3 311
GenBank mRNA BC061529.1 - 3 312
Y15068.1 - 2 246

View Full Product Details